2-Bromomethyl-1,3,5-trimethylbenzene 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)-1,3,5-trimethylbenzene

A321511-1G A321511-5G
103825-278EA 78 USD
103825-278 103836-212 76839-118 77306-242
2-Bromomethyl-1,3,5-trimethylbenzene 98%
2-Bromomethyl-1,3,5-trimethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
MW: 213.114 g/mol
Boiling Pt: 256.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09056829
CAS Number: 4761-00-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR