2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide 95%

Supplier: Ambeed
Danger

A101545-100G A101545-10G A101545-250MG A101545-25G A101545-1G A101545-5G
103796-020EA 10 USD
103796-020 103821-704 77135-504 77135-506 77135-508 77135-510 77297-144 77297-146
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide 95%
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
Melting Pt: 205 °C
Flash Pt: 104.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02181196
CAS Number: 17570-98-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR