Bis-TRIS hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol hydrochloride, 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane hydrochloride

A175351-25G A175351-100G
103845-338EA 9 USD
103845-338 76690-334 77082-680 77877-253
Bis-TRIS hydrochloride 97%
Bis-TRIS hydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₀NO₅Cl
Boiling Pt: 484.2 °C (760 mm Hg)
Flash Pt: 246.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00171252
CAS Number: 124763-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR