2-(Benzyloxy)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: Ethylene Glycol Monobenzyl Ether, Benzyl Cellosolve, Benzyl Glycol

A100024-100G A100024-25G A100024-500G A100024-5G A100024-1000G
103807-744EA 87 USD
103807-744 77245-034 77245-036 77245-038 77245-040 77296-540
2-(Benzyloxy)ethanol 98%
2-(Benzyloxy)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 254.4 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002868
CAS Number: 622-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR