2-(Benzyloxy)-1-ethanamine 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Benzyloxy)ethanamine, 2-(Benzyloxy)ethylamine

A192078-1G A192078-5G A192078-10G
103816-414EA 11 USD
103816-414 103847-018 77084-888 77084-890 77550-234
2-(Benzyloxy)-1-ethanamine 98%
2-(Benzyloxy)-1-ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.2056 g/mol
Boiling Pt: 240.6 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00235180
CAS Number: 38336-04-8
UN: 3267
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR