Bzl-Gly-OH 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzylamino)acetic acid, N-Benzylglycine

A155349-100G A155349-10G A155349-25G
103842-944EA 9 USD
103842-944 76871-824 76871-826 77318-662
Bzl-Gly-OH 98%
Bzl-Gly-OH

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 308.9 °C (760 mm Hg)
Melting Pt: 232 °C
Flash Pt: 140.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190772
CAS Number: 17136-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR