2-(Benzyl(tert-butoxycarbonyl)amino)acetic acid 97%

Supplier: Ambeed
Warning

A146117-250MG A146117-5G
103803-630EA 39 USD
103803-630 103841-716 77027-902 77317-024
2-(Benzyl(tert-butoxycarbonyl)amino)acetic acid 97%
2-(Benzyl(tert-butoxycarbonyl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
Boiling Pt: 409.491 °C (760 mm Hg)
Flash Pt: 201.453 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094473
CAS Number: 76315-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR