5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(benzofuran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Benzofuran, 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran
A327970-250MG
A327970-5G
103825-486EA
19
USD
103825-486
103836-438
76773-050
77306-366
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan 97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Precaución: for Research Use Only
Formula:
C₁₄H₁₇BO₃ MW: 244.1 g/mol Boiling Pt: 340 °C (760 mm Hg) Melting Pt: 72 °C Flash Pt: 159.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04115374 CAS Number: 519054-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...