2-(Benzo[d][1,3]dioxol-5-ylamino)ethanol hydrochloride 95%

Supplier: Ambeed
Warning

A197443-1G A197443-250MG
103816-924EA 10 USD
103816-924 103847-546 77085-558 77237-074
2-(Benzo[d][1,3]dioxol-5-ylamino)ethanol hydrochloride 95%
2-(Benzo[d][1,3]dioxol-5-ylamino)ethanol hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₃
MW: 217.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07779259
CAS Number: 94158-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR