2-Azetidin-3-yl-propan-2-ol hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Azetidin-3-yl)propan-2-ol hydrochloride

A334827-250MG A334827-1G A334827-5G
103825-666EA 98 USD
103825-666 103836-636 77167-470 77306-468 77306-470
2-Azetidin-3-yl-propan-2-ol hydrochloride 95%
2-Azetidin-3-yl-propan-2-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22199153
CAS Number: 1357923-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR