2-(Aminooxy)ethanol 98%

Supplier: Ambeed
Danger

A134666-250MG A134666-5G A134666-1G A134666-100MG
103802-036EA 88 USD
103802-036 76811-440 77298-716 77314-812 77599-946
2-(Aminooxy)ethanol 98%
2-(Aminooxy)ethanol

Precaución: for Research Use Only

Formula: C₂H₇NO₂
Boiling Pt: 238.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00517034
CAS Number: 3279-95-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR