2-(Aminomethyl)benzonitrile hydrochloride 97%

Supplier: Ambeed
Warning

A356657-1G A356657-250MG A356657-100MG
103826-176EA 44 USD
103826-176 103837-182 76840-324 77672-788 77672-786
2-(Aminomethyl)benzonitrile hydrochloride 97%
2-(Aminomethyl)benzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD06797973
CAS Number: 1134529-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR