2-(Aminomethyl)-5-bromophenol 95%

Supplier: Ambeed
Warning

A220234-100MG A220234-250MG
103818-844EA 413 USD
103818-844 103849-726 77165-184 77165-186
2-(Aminomethyl)-5-bromophenol 95%
2-(Aminomethyl)-5-bromophenol

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 305.8±27.0 °C (760 mm Hg)
Flash Pt: 138.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09999926
CAS Number: 1097778-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR