2-(Aminomethyl)-4-bromophenol hydrochloride 97%

Supplier: Ambeed
Warning

A230057-1G A230057-250MG
103819-516EA 82 USD
103819-516 103850-416 77089-268 77261-538
2-(Aminomethyl)-4-bromophenol hydrochloride 97%
2-(Aminomethyl)-4-bromophenol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉BrClNO
MW: 238.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045214
CAS Number: 1803612-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR