1-Adamantaneacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane

A139949-100G A139949-10G A139949-1G A139949-25G A139949-5G
103802-798EA 12 USD
103802-798 76719-322 76719-324 77299-100 77315-904 77315-906
1-Adamantaneacetic acid 98%
1-Adamantaneacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₂
Boiling Pt: 319.2 °C (760 mm Hg)
Melting Pt: 136…138 °C
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00074728
CAS Number: 4942-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR