9-Oxoacridine-10-acetic acid (CMA) 99+%

Supplier: Ambeed
Warning

Synonyms: Cridanimod, 2-(9-Oxoacridin-10(9H)-yl)acetic acid, 10-(Carboxymethyl)-9(10H)-acridone, 9-Oxo-10(9H)-acridineacetic Acid, 10-carboxymethyl-9-acridanone

A573261-1G A573261-5G
103828-814EA 14 USD
103828-814 103839-956 76844-086 76844-088
9-Oxoacridine-10-acetic acid (CMA) 99+%
9-Oxoacridine-10-acetic acid (CMA)

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO₃
Boiling Pt: 486.6 °C (760 mm Hg)
Melting Pt: ~289 °C
Flash Pt: 248.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00157050
CAS Number: 38609-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR