2-(9H-Carbazol-9-yl)ethanol 95%

Supplier: Ambeed
Warning

A135374-1G A135374-5G
103802-142EA 23 USD
103802-142 77298-776 77314-960
2-(9H-Carbazol-9-yl)ethanol 95%

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00518704
CAS Number: 1484-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR