2-(7-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-oxo-2H-chromen-4-yl)acetic acid 95%

Supplier: Ambeed
Warning

A102229-100MG A102229-1G
103796-340EA 443 USD
103796-340 76780-528 77282-916
2-(7-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-oxo-2H-chromen-4-yl)acetic acid 95%
2-(7-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-oxo-2H-chromen-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₂₆H₁₉NO₆
MW: 441.43 g/mol
Boiling Pt: 660.549 °C (760 mm Hg)
Flash Pt: 353.288 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13195295
CAS Number: 378247-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR