(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid 98%

Supplier: Ambeed
Warning

A152209-100G A152209-1G A152209-25G A152209-5G
103804-364EA 13 USD
103804-364 103842-526 76783-414 77318-138 77318-140 77318-142
(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid 98%
(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₃
Boiling Pt: 403.9 °C (760 mm Hg)
Melting Pt: 157 °C
Flash Pt: 154.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00439456
CAS Number: 23981-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR