6-Chloro-3-pyridineacetic acid 98%
Supplier: Ambeed
|
Warning
|
A223950-100G
A223950-10G
A223950-1G
A223950-25G
A223950-5G
A223950-500G
103819-074EA
5
USD
103819-074
103849-958
76975-912
76975-914
77088-588
77088-590
77303-678
77881-568
6-Chloro-3-pyridineacetic acid 98%
6-Chloro-3-pyridineacetic acid
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ Boiling Pt: 336.6 °C (760 mm Hg) Melting Pt: 164…169 °C Flash Pt: 157.378 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01863172 CAS Number: 39891-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...