2-(6-Chloro-1H-indol-3-yl)acetic acid 97%

Supplier: Ambeed
Warning

A287515-100MG A287515-250MG
103813-760EA 21 USD
103813-760 103834-902 77032-236 77032-238
2-(6-Chloro-1H-indol-3-yl)acetic acid 97%
2-(6-Chloro-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MW: 209.62 g/mol
Boiling Pt: 445.6 °C (760 mm Hg)
Flash Pt: 223.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09751729
CAS Number: 1912-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR