5-Hydroxyindol-3-ylacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Hydroxyindole-3-acetic acid, 2-(5-Hydroxy-1H-indol-3-yl)acetic acid
A145064-1MG
A145064-100MG
A145064-250MG
103803-490EA
67
USD
103803-490
103841-572
76798-408
77600-084
77600-086
5-Hydroxyindol-3-ylacetic acid 98%
5-Hydroxyindol-3-ylacetic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol Boiling Pt: 497.1 °C (760 mm Hg) Melting Pt: 161…164 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005639 CAS Number: 54-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...