2-(5-Chloropyridin-2-yl)acetonitrile 95%

Supplier: Ambeed
Warning

A116578-100MG A116578-250MG A116578-5G A116578-1G
103798-758EA 75 USD
103798-758 103824-622 76810-218 76810-220 77297-476 77311-426
2-(5-Chloropyridin-2-yl)acetonitrile 95%
2-(5-Chloropyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09923938
CAS Number: 185315-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR