2-(5-Chloro-1H-indazol-3-yl)acetic acid 97%

Supplier: Ambeed
Warning

A121995-250MG A121995-1G
103800-168EA 129 USD
103800-168 76870-256 77297-854
2-(5-Chloro-1H-indazol-3-yl)acetic acid 97%
2-(5-Chloro-1H-indazol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
Boiling Pt: 463.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09835467
CAS Number: 27328-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR