2-(5-Bromopyrimidin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A378410-1G A378410-5G A378410-250MG
103826-624EA 54 USD
103826-624 103837-658 77306-902 77306-904 77572-124
2-(5-Bromopyrimidin-2-yl)acetic acid 95%
2-(5-Bromopyrimidin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.0201 g/mol
Boiling Pt: 349 °C (760 mm Hg)
Flash Pt: 164.9 °C
Storage Temperature: Freezer
MDL Number: MFCD09909705
CAS Number: 1134327-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR