2-(5-Bromopyridin-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A259434-1G A259434-25G A259434-5G A259434-100MG A259434-10G A259434-250MG
103812-442EA 25 USD
103812-442 103852-322 77091-884 77091-886 77091-888 77113-658 77113-660 77113-662
2-(5-Bromopyridin-2-yl)acetonitrile 97%
2-(5-Bromopyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
Melting Pt: 29 °C
Flash Pt: 126.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07644625
CAS Number: 312325-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR