2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 97%

Supplier: Ambeed
Warning

A173680-1G A173680-10G
103806-856EA 10 USD
103806-856 103845-156 77082-424 77138-490
2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 97%
2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄BNO₂
MW: 215.05 g/mol
Boiling Pt: 368.1 °C (760 mm Hg)
Flash Pt: 176.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039011
CAS Number: 214360-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR