N-(4-Oxocyclohexyl)phthalimide 97%

Supplier: Ambeed
Warning

Synonyms: 4-Phthalimidocyclohexanone, 2-(4-Oxocyclohexyl)isoindoline-1,3-dione

A137967-1G A137967-5G
103802-536EA 10 USD
103802-536 77298-938 77315-536
N-(4-Oxocyclohexyl)phthalimide 97%
N-(4-Oxocyclohexyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₃
MW: 243.26 g/mol
Boiling Pt: 409.96 °C (760 mm Hg)
Flash Pt: 191.122 °C
Storage Temperature: Ambient
MDL Number: MFCD00158659
CAS Number: 104618-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR