2-(4-Methyl-1-piperazinyl)aniline 98%

Supplier: Ambeed
Warning

A825082-1G A825082-5G A825082-10G
103841-152EA 48 USD
103841-152 76845-388 76845-390 77006-706 77006-708
2-(4-Methyl-1-piperazinyl)aniline 98%
2-(4-Methyl-1-piperazinyl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃
Boiling Pt: 321.2 °C (760 mm Hg)
Melting Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD04035359
CAS Number: 180605-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR