(4-Methyl-1H-pyrazol-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

A125133-250MG A125133-5G
103800-628EA 19 USD
103800-628 76983-432 77298-034
(4-Methyl-1H-pyrazol-1-yl)acetic acid 95%
(4-Methyl-1H-pyrazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
Boiling Pt: 308.7±25.0 °C (760 mm Hg)
Flash Pt: 140.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419610
CAS Number: 956364-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR