2-(4-Methoxyphenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A391218-250MG A391218-1G
103826-876EA 139 USD
103826-876 103837-916 76817-774 76841-306
2-(4-Methoxyphenyl)propanoic acid 95%
2-(4-Methoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 309.4 °C (760 mm Hg)
Flash Pt: 121.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00969551
CAS Number: 942-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR