2-(4-Methoxyphenyl)dihydro-2H-pyran-4(3H)-one 96%

Supplier: Ambeed
Warning

A227073-250MG A227073-1G
103819-290EA 315 USD
103819-290 103850-198 76874-160 77303-788
2-(4-Methoxyphenyl)dihydro-2H-pyran-4(3H)-one 96%
2-(4-Methoxyphenyl)dihydro-2H-pyran-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19650726
CAS Number: 1026692-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR