2-(4-Methoxyphenoxy)-1-ethanamine 97%

Supplier: Ambeed
Danger

A706342-1G A706342-250MG A706342-5G
103840-668EA 105 USD
103840-668 76844-880 77243-530 77243-532 77308-172
2-(4-Methoxyphenoxy)-1-ethanamine 97%
2-(4-Methoxyphenoxy)-1-ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
Boiling Pt: 283.4 °C (760 mm Hg)
Melting Pt: 40…43 °C at N/A mmHg
Flash Pt: 136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052978
CAS Number: 50800-92-5
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR