2-(4-Iodophenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A364204-100G
A364204-10G
A364204-250MG
A364204-1G
A364204-25G
A364204-5G
103826-356EA
14
USD
103826-356
103837-374
76788-586
76788-588
76788-590
76840-600
76840-602
76840-604
2-(4-Iodophenyl)ethanol 98%
2-(4-Iodophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₉IO Boiling Pt: 275.7 °C (760 mm Hg) Melting Pt: 48…49 °C Flash Pt: 121 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173705 CAS Number: 52914-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...