(4-Iodophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile

A146276-100G A146276-10G A146276-250MG A146276-1G A146276-25G A146276-5G
103803-650EA 17 USD
103803-650 103841-736 77095-636 77095-638 77095-640 77317-050 77317-052 77317-054
(4-Iodophenyl)acetonitrile 98%
(4-Iodophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₆IN
Boiling Pt: 285.8 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060306
CAS Number: 51628-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR