2-(4-Hydroxyphenyl)ethanol 97%

Supplier: Ambeed
Warning

A124064-100G A124064-100MG A124064-25G A124064-25MG A124064-500G A124064-10G A124064-1000G
77189-798EA 53 USD
77189-798 77189-800 77189-802 77189-804 77189-806 77571-450 77745-910
2-(4-Hydroxyphenyl)ethanol 97%
2-(4-Hydroxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00002902
CAS Number: 501-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR