(4-Hydroxyphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxybenzyl cyanide, 2-(4-Hydroxyphenyl)acetonitrile

A190006-25G A190006-100G A190006-10G A190006-500G A190006-5G
103846-844EA 5 USD
103846-844 77084-638 77236-846 77236-848 77236-850 77302-196
(4-Hydroxyphenyl)acetonitrile 98%
(4-Hydroxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 330 °C (760 mm Hg)
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002383
CAS Number: 14191-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR