2-(4-Hydroxyphenyl)-2-methylpropanoic acid 97%

Supplier: Ambeed
Warning

A118061-100MG A118061-250MG A118061-5G A118061-1G
103798-982EA 120 USD
103798-982 76983-074 76983-076 76983-078 77297-578
2-(4-Hydroxyphenyl)-2-methylpropanoic acid 97%
2-(4-Hydroxyphenyl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Boiling Pt: 345.5 °C (760 mm Hg)
Flash Pt: 177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00995712
CAS Number: 29913-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR