2-(4-Hydroxybenzoyl)benzoic acid 98%

Supplier: Ambeed
Warning

A123510-10G A123510-25G A123510-5G
77271-654EA 10.3 USD
77271-654 77271-656 77297-946
2-(4-Hydroxybenzoyl)benzoic acid 98%
2-(4-Hydroxybenzoyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MW: 242.227 g/mol
Boiling Pt: 512.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00016507
CAS Number: 85-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR