2-(4-Fluorophenyl)ethanol 98%

Supplier: Ambeed
Warning

A203886-100G A203886-10G A203886-500G A203886-25G A203886-5G
103817-502EA 10 USD
103817-502 103848-162 76691-286 76691-288 76691-290 77086-302 77086-304
2-(4-Fluorophenyl)ethanol 98%
2-(4-Fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉FO
Boiling Pt: 213 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002898
CAS Number: 7589-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR