2-(4-Fluorophenyl)-5-(5-iodo-2-methylbenzyl)thiophene 98%

Supplier: Ambeed
Warning

A126498-100G A126498-250MG A126498-1G A126498-25G A126498-5G A126498-10G
103800-822EA 18 USD
103800-822 76782-050 76782-052 77313-040 77313-042 77313-044 77666-800
2-(4-Fluorophenyl)-5-(5-iodo-2-methylbenzyl)thiophene 98%
2-(4-Fluorophenyl)-5-(5-iodo-2-methylbenzyl)thiophene

Precaución: for Research Use Only

Formula: C₁₈H₁₄FIS
MW: 408.27 g/mol
Boiling Pt: 450.2 °C (760 mm Hg)
Melting Pt: 107…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12405593
CAS Number: 898566-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR