2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A156148-10G A156148-250MG A156148-25G A156148-1G A156148-5G
103804-834EA 15 USD
103804-834 103843-038 77096-000 77096-002 77096-004 77318-786 77318-788
2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine 97%
2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₄FN₃
MW: 327.36 g/mol
Boiling Pt: 537.4±52.0 °C (760 mm Hg)
Flash Pt: 278.8 °C
Storage Temperature: Ambient
MDL Number: MFCD28964710
CAS Number: 203450-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR