4-Fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluorophenylboronic acid Pinacol ester, 1-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

A165629-100G A165629-1G A165629-5G
103805-980EA 5 USD
103805-980 103844-236 76720-296 76720-298 77320-456 77320-458
4-Fluorophenylboronic acid pinacol ester 97%
4-Fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₂
Boiling Pt: 279.4 °C (760 mm Hg)
Flash Pt: 122.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03453665
CAS Number: 214360-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR