2-(4-Fluoro-3-nitrophenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A107559-100G
A107559-10G
A107559-25G
A107559-1G
A107559-5G
103797-214EA
9
USD
103797-214
103822-980
76780-904
76780-906
76780-908
77309-838
77309-840
2-(4-Fluoro-3-nitrophenyl)acetic acid 98%
2-(4-Fluoro-3-nitrophenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₆FNO₄ MW: 199.1400 g/mol Flash Pt: 190.397 °C Storage Temperature: Ambient |
MDL Number:
MFCD09743463 CAS Number: 192508-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...