2-(4-Fluoro-3-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

A175255-100MG A175255-250MG
103807-018EA 286 USD
103807-018 103845-322 77260-064 77260-066
2-(4-Fluoro-3-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(4-Fluoro-3-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₂BFO₂
MW: 264.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18732290
CAS Number: 2828446-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR