(4-Fluoro-2-methylphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Fluoro-2-methylphenyl)acetic acid

A226419-250MG A226419-1G
103819-244EA 40 USD
103819-244 103850-146 76786-132 77088-876
(4-Fluoro-2-methylphenyl)acetic acid 97%
(4-Fluoro-2-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00049318
CAS Number: 407640-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR