4-Cyanophenylacetic acid 98%

Supplier: Ambeed
Warning

A207856-1G A207856-5G A207856-100G A207856-10G A207856-250MG A207856-25G
103817-834EA 10 USD
103817-834 103848-520 77086-814 77086-816 77164-864 77164-866 77164-868 77303-048
4-Cyanophenylacetic acid 98%
4-Cyanophenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
Boiling Pt: 355.2 °C (760 mm Hg)
Melting Pt: 150…154 °C
Flash Pt: 168.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06798066
CAS Number: 5462-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR