2-(4-Chlorophenyl)cyclopropanecarboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A174238-100MG
A174238-250MG
A174238-1G
103806-934EA
111
USD
103806-934
103845-232
76998-326
76998-328
77082-528
2-(4-Chlorophenyl)cyclopropanecarboxylic acid 95%
2-(4-Chlorophenyl)cyclopropanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉ClO₂ Boiling Pt: 346.9 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD03419437 CAS Number: 90940-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...