(4-Chlorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

A119312-100G A119312-25G A119312-500G
103808-204EA 57 USD
103808-204 76995-724 76995-726 76995-728
(4-Chlorophenyl)acetic acid 98%
(4-Chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 294.1 °C (760 mm Hg)
Melting Pt: 102…105 °C
Flash Pt: 131.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR