2-(4-Chlorophenyl)acetamide 95%

Supplier: Ambeed
Warning

A211604-1G A211604-25G A211604-5G A211604-100MG A211604-10G A211604-250MG
103818-120EA 35 USD
103818-120 103848-814 77087-232 77087-234 77087-236 77097-688 77097-690 77097-692
2-(4-Chlorophenyl)acetamide 95%
2-(4-Chlorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 344.9±25.0 °C (760 mm Hg)
Melting Pt: 183…185 °C
Flash Pt: 162.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02380705
CAS Number: 20101-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR